Assay Details
Gene Symbol: gly-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059473.8 | NP_491874.1 | 4 - 5 | 579 | 134 | |
GenBank mRNA | AF154122.1 | - | 4 - 5 | 579 | 134 | |
AY037802.1 | - | 8 - 9 | 962 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
