Assay Details
Gene Symbol: unc-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026700.2 | NP_001021871.1 | 15 - 16 | 2017 | 78 | |
NM_001026702.3 | NP_001021873.1 | 17 - 18 | 3043 | 78 | ||
NM_001026703.4 | NP_001021874.1 | 16 - 17 | 2026 | 78 | ||
NM_001263573.3 | NP_001250502.1 | 16 - 17 | 2035 | 78 | ||
NM_001306224.3 | NP_001293153.1 | 4 - 5 | 1115 | 78 | ||
NM_001330927.3 | NP_001317864.1 | 15 - 16 | 2026 | 78 | ||
NM_001330928.3 | NP_001317865.1 | 15 - 16 | 1981 | 78 | ||
NM_001330929.3 | NP_001317866.1 | 15 - 16 | 1972 | 78 | ||
NM_001330930.3 | NP_001317863.1 | 7 - 8 | 994 | 78 | ||
NM_001330931.3 | NP_001317867.1 | 3 - 4 | 973 | 78 | ||
NM_001380703.1 | NP_001367037.1 | 8 - 9 | 1003 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
