Assay Details
Gene Symbol: unc-13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026700.2 | NP_001021871.1 | 21 - 22 | 3356 | 93 | |
NM_001026702.3 | NP_001021873.1 | 23 - 24 | 4382 | 93 | ||
NM_001026703.4 | NP_001021874.1 | 22 - 23 | 3365 | 93 | ||
NM_001263573.3 | NP_001250502.1 | 22 - 23 | 3374 | 93 | ||
NM_001306224.3 | NP_001293153.1 | 10 - 11 | 2454 | 93 | ||
NM_001330927.3 | NP_001317864.1 | 21 - 22 | 3365 | 93 | ||
NM_001330928.3 | NP_001317865.1 | 21 - 22 | 3320 | 93 | ||
NM_001330929.3 | NP_001317866.1 | 21 - 22 | 3311 | 93 | ||
NM_001330930.3 | NP_001317863.1 | 13 - 14 | 2333 | 93 | ||
NM_001330931.3 | NP_001317867.1 | 9 - 10 | 2312 | 93 | ||
NM_001380703.1 | NP_001367037.1 | 14 - 15 | 2342 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
