Assay Details
Gene Symbol: rme-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026224.6 | NP_001021395.1 | 11 - 12 | 3937 | 113 | |
NM_059821.5 | NP_492222.2 | 11 - 12 | 3930 | 113 | ||
GenBank mRNA | AF372457.1 | - | 11 - 12 | 3930 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
