Assay Details
Gene Symbol: rme-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026224.6 | NP_001021395.1 | 21 - 22 | 6750 | 84 | |
NM_059821.5 | NP_492222.2 | 22 - 23 | 6767 | 84 | ||
GenBank mRNA | AF372457.1 | - | 22 - 23 | 6767 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
