Assay Details
Gene Symbol: ppfr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026217.4 | NP_001021388.1 | 15 - 16 | 3163 | 109 | |
NM_001026218.5 | NP_001021389.1 | 14 - 15 | 2791 | 109 | ||
NM_001128984.4 | NP_001122456.2 | 17 - 18 | 3330 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
