Assay Details
Gene Symbol: ppfr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026217.4 | NP_001021388.1 | 3 - 4 | 390 | 77 | |
NM_001026218.5 | NP_001021389.1 | 3 - 4 | 396 | 77 | ||
NM_001026219.5 | NP_001021390.1 | 3 - 4 | 341 | 77 | ||
NM_001128984.4 | NP_001122456.2 | 5 - 6 | 557 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
