Assay Details
Gene Symbol: Y119C1A.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059013.5 | NP_491414.3 | 3 - 4 | 571 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Y119C1A.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059013.5 | NP_491414.3 | 3 - 4 | 571 | 123 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |