Assay Details
Gene Symbol: max-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380794.1 | NP_001367137.1 | 8 - 9 | 2187 | 79 | |
NM_001383435.1 | 9 - 10 | 2329 | 79 | |||
GenBank mRNA | AY084080.1 | - | 8 - 9 | 2187 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
