Assay Details
Gene Symbol: F10G7.10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001381399.2 | NP_001366635.1 | 14 - 15 | 3080 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
