Assay Details
Gene Symbol: cks-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380372.2 | NP_001367486.1 | 6 - 7 | 1323 | 97 |
Gene Symbol: C09G4.4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380371.2 | NP_001367487.1 | 6 - 7 | 1371 | 97 |
Gene Symbol: mes-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383148.1 | 6 - 7 | 1382 | 97 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001380371.2, NM_001380372.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
