Assay Details
Gene Symbol: pqn-85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001377684.3 | NP_001364775.1 | 30 - 31 | 7250 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
