Assay Details
Gene Symbol: pqn-85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313675.3 | NP_001300604.1 | 3 | 657 | 76 | |
NM_001377684.3 | NP_001364775.1 | 33 - 34 | 8038 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
