Assay Details
Gene Symbol: pqn-85 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313675.3 | NP_001300604.1 | 5 | 1103 | 73 | |
NM_001377684.3 | NP_001364775.1 | 35 - 36 | 8484 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
