Assay Details
Gene Symbol: B0432.7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001313561.2 | NP_001300490.1 | 2 - 3 | 348 | 113 | |
NM_001313562.3 | NP_001300491.1 | 1 - 2 | 101 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
