Assay Details
Gene Symbol: gcp-2.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_075684.7 | NP_508085.2 | 1 - 2 | 50 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
