Assay Details
Gene Symbol: tpp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_062820.5 | NP_495221.1 | 7 - 8 | 3058 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
