Assay Details
Gene Symbol: C16D2.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_063547.5 | NP_495948.1 | 3 - 4 | 1013 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
