Assay Details
Gene Symbol: acl-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_064324.9 | NP_496725.2 | 2 - 3 | 500 | 59 | |
GenBank mRNA | AJ414404.1 | - | 2 - 3 | 495 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
