Assay Details
Gene Symbol: egl-45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_066297.7 | NP_498698.1 | 3 - 4 | 689 | 147 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
