Assay Details
Gene Symbol: T05G5.5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268094.3 | NP_001255023.1 | 8 - 9 | 1509 | 70 | |
NM_001268095.3 | NP_001255024.1 | 3 - 4 | 386 | 70 | ||
NM_001268096.3 | NP_001255025.1 | 3 - 4 | 329 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
