Assay Details
Gene Symbol: T05G5.5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268094.3 | NP_001255023.1 | 9 - 10 | 1673 | 73 | |
NM_001268095.3 | NP_001255024.1 | 4 - 5 | 550 | 73 | ||
NM_001268096.3 | NP_001255025.1 | 4 - 5 | 493 | 73 | ||
NM_001268097.3 | NP_001255026.1 | 1 - 2 | 61 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
