Assay Details
Gene Symbol: T05G5.5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268094.3 | NP_001255023.1 | 2 - 3 | 525 | 102 |
Gene Symbol: ech-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379869.2 | NP_001366674.1 | 2 - 3 | 564 | 102 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001268094.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
