Assay Details
Gene Symbol: gbf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001379938.1 | NP_001366879.1 | 6 - 7 | 1104 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
