Assay Details
Gene Symbol: F35H10.10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068999.7 | NP_501400.1 | 3 - 4 | 709 | 87 | |
GenBank mRNA | U70870.1 | - | 3 - 4 | 702 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
