Assay Details
Gene Symbol: sri-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276854.1 | NP_001263783.1 | 3 - 4 | 364 | 95 | |
NM_069642.1 | NP_502043.1 | 3 - 4 | 517 | 95 | ||
NR_131607.1 | 4 - 5 | 550 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
