Assay Details
Gene Symbol: ZK829.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069863.5 | NP_502264.2 | 1 - 2 | 335 | 114 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
