Assay Details
Gene Symbol: unc-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268672.3 | NP_001255601.1 | 10 - 11 | 1648 | 72 | |
NM_001268673.3 | NP_001255602.1 | 10 - 11 | 1648 | 72 | ||
NM_001307045.3 | NP_001293974.1 | 10 - 11 | 1648 | 72 | ||
NM_001307046.3 | NP_001293975.1 | 10 - 11 | 1648 | 72 | ||
NM_069872.6 | NP_502273.2 | 10 - 11 | 1775 | 72 | ||
NM_069873.6 | NP_502274.2 | 10 - 11 | 1648 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
