Assay Details
Gene Symbol: grd-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307786.2 | NP_001294715.1 | 5 - 6 | 878 | 66 | |
NM_001368462.3 | NP_001355524.1 | 5 - 6 | 1000 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
