Assay Details
Gene Symbol: wdr-20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001348580.3 | NP_001335505.1 | 10 | 1925 | 112 | |
NM_001380736.2 | NP_001368633.1 | 8 - 9 | 1808 | 112 | ||
NM_001383378.1 | 9 - 10 | 1923 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
