Assay Details
Gene Symbol: nrx-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001269333.4 | NP_001256262.1 | 23 - 24 | 4104 | 106 | |
NM_001269335.3 | NP_001256264.1 | 3 - 4 | 173 | 106 | ||
NM_001276931.3 | NP_001263860.1 | 24 - 25 | 4118 | 106 | ||
NM_001307745.4 | NP_001294674.1 | 2 - 3 | 528 | 106 | ||
GenBank mRNA | KF227795.1 | - | 2 - 3 | 332 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
