Assay Details
Gene Symbol: par-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028847.4 | NP_001024018.1 | 3 - 4 | 199 | 87 | |
NM_001269631.3 | NP_001256560.1 | 3 - 4 | 199 | 87 | ||
NM_001380808.2 | NP_001368670.1 | 3 - 4 | 496 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
