Assay Details
Gene Symbol: grd-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074281.3 | NP_506682.1 | 1 - 2 | 87 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: grd-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_074281.3 | NP_506682.1 | 1 - 2 | 87 | 67 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |