Assay Details
Gene Symbol: rgl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076149.7 | NP_508550.1 | 1 - 2 | 176 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
