Assay Details
Gene Symbol: rgl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129668.5 | NP_001123140.1 | 9 - 10 | 2094 | 84 | |
NM_076149.7 | NP_508550.1 | 8 - 9 | 2108 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
