Assay Details
Gene Symbol: acn-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029282.6 | NP_001024453.1 | 3 - 4 | 587 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
