Assay Details
Gene Symbol: grd-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076823.5 | NP_509224.4 | 4 - 5 | 1056 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
