Assay Details
Gene Symbol: gst-42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077561.8 | NP_509962.1 | 3 - 4 | 447 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
