Assay Details
Gene Symbol: lsd-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077599.6 | NP_510000.2 | 8 - 9 | 1737 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
