Assay Details
Gene Symbol: lyst-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_077629.4 | NP_510030.2 | 1 - 2 | 89 | 170 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
