Assay Details
Gene Symbol: ENSA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005630758.3 | XP_005630815.1 | 2 - 3 | 228 | 71 | |
XM_022404971.1 | XP_022260679.1 | 2 - 3 | 159 | 71 | ||
XM_855901.5 | XP_860994.1 | 2 - 3 | 334 | 71 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
