Assay Details
Gene Symbol: RNF128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003435638.3 | XP_003435686.1 | 5 - 6 | 1144 | 75 | |
XM_549171.6 | XP_549171.3 | 5 - 6 | 1221 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
