Assay Details
Gene Symbol: CPEB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005619196.3 | XP_005619253.1 | 10 - 11 | 3221 | 67 | |
XM_005619197.3 | XP_005619254.1 | 9 - 10 | 3197 | 67 | ||
XM_005619198.3 | XP_005619255.1 | 8 - 9 | 3158 | 67 | ||
XM_005619199.3 | XP_005619256.1 | 7 - 8 | 3107 | 67 | ||
XM_847725.5 | XP_852818.1 | 9 - 10 | 1960 | 67 | ||
XM_861280.5 | XP_866373.1 | 8 - 9 | 3131 | 67 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
