Assay Details
Gene Symbol: QKI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003021.2 | NP_001003021.1 | 418 | 80 | ||
GenBank mRNA | AB047312.1 | - | 422 | 80 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
