Assay Details
Gene Symbol: CYP1A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008720.1 | NP_001008720.1 | 2 - 3 | 878 | 70 | |
GenBank mRNA | AB094347.1 | - | 2 - 3 | 878 | 70 | |
AB094348.1 | - | 2 - 3 | 878 | 70 | ||
AB094349.1 | - | 2 - 3 | 878 | 70 | ||
AB159771.1 | - | 2 - 3 | 878 | 70 | ||
HG965165.1 | - | 1 - 2 | 816 | 70 | ||
HG965167.1 | - | 1 - 2 | 816 | 70 | ||
HG974425.1 | - | 1 - 2 | 816 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
