Assay Details
Gene Symbol: EDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002956.1 | NP_001002956.1 | 4 - 5 | 680 | 64 | |
GenBank mRNA | AB115087.1 | - | 4 - 5 | 680 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
