Assay Details
Gene Symbol: RARA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012645.1 | NP_001012663.1 | 170 | 91 | ||
GenBank mRNA | AB179779.1 | - | 170 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001012645.1 transcripts.
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
