Assay Details
Gene Symbol: RARA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012645.1 | NP_001012663.1 | 717 | 70 | ||
XM_005624336.3 | XP_005624393.1 | 695 | 70 | |||
GenBank mRNA | AB179779.1 | - | 717 | 70 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
