Assay Details
Gene Symbol: RARA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012645.1 | NP_001012663.1 | 1551 | 73 | ||
XM_005624336.3 | XP_005624393.1 | 1529 | 73 | |||
XM_005624338.2 | XP_005624395.1 | 901 | 73 | |||
XM_005624339.2 | XP_005624396.1 | 889 | 73 | |||
XM_022422575.1 | XP_022278283.1 | 871 | 73 | |||
XM_022422576.1 | XP_022278284.1 | 1054 | 73 | |||
GenBank mRNA | AB179779.1 | - | 1551 | 73 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
