Assay Details
Gene Symbol: COMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004074.1 | NP_001004074.1 | 1 - 2 | 212 | 53 | |
XM_005636532.3 | XP_005636589.1 | 2 - 3 | 405 | 53 | ||
XM_005636533.3 | XP_005636590.1 | 2 - 3 | 417 | 53 | ||
XM_014108109.2 | XP_013963584.1 | 2 - 3 | 653 | 53 | ||
GenBank mRNA | AB193089.1 | - | 1 - 2 | 139 | 53 | |
AY340812.1 | - | 1 - 2 | 212 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
